Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 8 | 1158 | ||
NM_001145103.1 | NP_001138575.1 | 8 | 969 | |||
NM_001145104.1 | NP_001138576.1 | 6 | 602 | |||
NM_005902.3 | NP_005893.1 | 8 | 1393 | |||
XM_011521559.2 | XP_011519861.1 | 7 | 1420 | |||
XM_011521560.1 | XP_011519862.1 | 8 | 1332 | |||
GenBank mRNA | AB209616.1 | - | 8 | 978 | ||
AK026690.1 | - | 1 | 791 | |||
AK290881.1 | - | 9 | 1318 | |||
AK298139.1 | - | 8 | 969 | |||
AK300614.1 | - | 6 | 599 | |||
AK301348.1 | - | 6 | 486 | |||
AK316017.1 | - | 8 | 1018 | |||
BC050743.1 | - | 8 | 1311 | 6094328 | ||
DA755047.1 | - | 3 | 443 | |||
U68019.1 | - | 8 | 1161 | |||
U76622.1 | - | 8 | 1132 |