Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001419.2 | NP_001001419.1 | 6 | 1145 | ||
NM_001001420.2 | NP_001001420.1 | 4 | 986 | |||
NM_005903.6 | NP_005894.3 | 5 | 1061 | |||
XM_017009470.1 | XP_016864959.1 | 6 | 1277 | |||
GenBank mRNA | AF009678.1 | - | 3 | 713 | ||
AF010601.1 | - | 3 | 830 | |||
AK313397.1 | - | 5 | 1061 | |||
BC009682.2 | - | 3 | 867 | 3906006 | ||
BX648328.1 | - | 5 | 1000 | |||
CR457424.1 | - | 3 | 699 | |||
CR749366.1 | - | 5 | 1000 | |||
CR749473.1 | - | 5 | 988 | |||
U73825.1 | - | 3 | 848 |