Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025839.3 | NP_080115.1 | 3 | 496 | ||
GenBank mRNA | AF435792.1 | - | 3 | 508 | ||
AK002378.1 | - | 3 | 489 | |||
AK003241.1 | - | 3 | 497 | |||
AK007897.1 | - | 3 | 492 | |||
AK019936.1 | - | 3 | 499 | |||
AK159841.1 | - | 3 | 489 | |||
AK186853.1 | - | 2 | 206 | |||
BC025569.1 | - | 3 | 570 | 5322456 | ||
BC028494.1 | - | 3 | 489 | 1511165 |