Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190821.1 | NP_001177750.1 | 1 | 836 | ||
NM_005904.3 | NP_005895.1 | 1 | 836 | |||
GenBank mRNA | AF010193.1 | - | 1 | 844 | ||
AF015261.1 | - | 1 | 549 | |||
BC074818.2 | - | 1 | 583 | 30915321 | ||
BC074819.2 | - | 1 | 583 | 30915535 | ||
DA882147.1 | - | 1 | 268 |