Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cir1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025854.3 | NP_080130.2 | 10 | 1098 | ||
GenBank mRNA | AK005900.1 | - | 4 | 591 | ||
AK006260.1 | - | 10 | 1010 | |||
AK008242.1 | - | 10 | 1062 | |||
AK131674.1 | - | 10 | 1074 |