Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cir1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025854.3 | NP_080130.2 | 8, 9 | 702 | ||
GenBank mRNA | AK005900.1 | - | 2, 3 | 195 | ||
AK006260.1 | - | 8, 9 | 614 | |||
AK008242.1 | - | 8, 9 | 666 | |||
AK131674.1 | - | 8, 9 | 678 | |||
BC012219.1 | - | 8, 9 | 811 | |||
BC061077.1 | - | 8, 9 | 637 | |||
BC089602.1 | - | 8, 9 | 650 | |||
BC094283.1 | - | 8, 9 | 638 | |||
BC099444.1 | - | 7, 8 | 1015 |