Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025862.2 | NP_080138.2 | 7 | 783 | ||
GenBank mRNA | AK009379.1 | - | 7 | 762 | ||
AK019502.1 | - | 7 | 780 | |||
AK029897.1 | - | 7 | 747 | |||
AK049908.1 | - | 3 | 1103 | |||
AK051091.1 | - | 7 | 738 | |||
AK077335.1 | - | 7 | 763 | |||
AK161255.1 | - | 7 | 737 | |||
AK178437.1 | - | 4 | 319 | |||
AK207169.1 | - | 4 | 271 | |||
BC026535.1 | - | 7 | 757 | |||
BC037644.1 | - | 7 | 759 | 5053565 | ||
BQ931973.1 | - | 7 | 783 |