Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025873.2 | NP_080149.2 | 7 | 914 | ||
GenBank mRNA | AK003556.1 | - | 7 | 915 | ||
AK010176.1 | - | 7 | 914 | |||
AK128921.1 | - | 7 | 913 | |||
BC019812.1 | - | 7 | 883 | 5042856 | ||
BC051040.1 | - | 7 | 890 |