Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024186.5 | NP_077148.1 | 2 | 206 | ||
NM_024272.4 | NP_077234.2 | 2 | 206 | |||
GenBank mRNA | AK005150.1 | - | 2 | 141 | ||
AK010220.1 | - | 2 | 148 | |||
AK013177.1 | - | 2 | 132 | |||
AK039140.1 | - | 998 | ||||
AK051840.1 | - | 2 | 143 | |||
AK133699.1 | - | 2 | 150 | |||
AY037837.1 | - | 2 | 162 | |||
AY167988.1 | - | 2 | 116 | |||
BC002176.1 | - | 2 | 218 | 3486988 |