Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025876.2 | NP_080152.1 | 12, 13 | 1596 | ||
GenBank mRNA | AK010081.1 | - | 12, 13 | 1596 | ||
AK010097.1 | - | 12, 13 | 1596 | |||
AK088517.1 | - | 12, 13 | 1605 | |||
AK210159.1 | - | 92 | ||||
BC025132.1 | - | 11, 12 | 1561 | 3666341 |