Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025881.3 | NP_080157.1 | 6, 7 | 801 | ||
NM_028190.3 | NP_082466.2 | 6, 7 | 801 | |||
GenBank mRNA | AK010539.1 | - | 6, 7 | 753 | ||
AK160293.1 | - | 6, 7 | 802 | |||
BC019194.1 | - | 7, 8 | 794 | |||
BC034135.1 | - | 5, 6 | 967 | 5003664 | ||
BC055875.1 | - | 6, 7 | 1784 | |||
BQ572015.1 | - | 4, 5 | 453 | |||
CF951283.1 | - | 2, 3 | 197 |