Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pole4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025882.3 | NP_080158.1 | 4 | 921 | ||
GenBank mRNA | AK010135.1 | - | 4 | 916 | ||
AK010300.1 | - | 4 | 922 | |||
AK167631.1 | - | 4 | 911 | |||
AK207330.1 | - | 1 | 183 | |||
BC023189.1 | - | 4 | 899 | 4952949 |