Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199216.1 | NP_001186145.1 | 4 | 359 | ||
NM_002361.3 | NP_002352.1 | 4 | 372 | |||
NM_080600.2 | NP_542167.1 | 4 | 372 | |||
GenBank mRNA | AK222649.1 | - | 4 | 297 | ||
AK222680.1 | - | 4 | 275 | |||
AK223562.1 | - | 4 | 333 | |||
AK294644.1 | - | 4 | 266 | |||
AK308976.1 | - | 4 | 284 | |||
BC053347.1 | - | 4 | 264 | 5177296 | ||
BC093045.1 | - | 4 | 324 | 30396005 | ||
BI755065.1 | - | 4 | 322 | |||
DC331168.1 | - | 4 | 372 | |||
M29273.1 | - | 4 | 294 |