Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033573.1 | NP_001028745.1 | 10, 11 | 1701 | ||
NM_025883.3 | NP_080159.3 | 10, 11 | 1585 | |||
GenBank mRNA | AK010296.1 | - | 10, 11 | 1580 | ||
AK011373.1 | - | 10, 11 | 1552 | |||
AK035172.1 | - | 1664 | ||||
AK050388.1 | - | 10, 11 | 1700 | |||
AK077693.1 | - | 10, 11 | 1539 | |||
AK144128.1 | - | 10, 11 | 1557 | |||
AK154741.1 | - | 10, 11 | 1563 | |||
AK157494.1 | - | 10, 11 | 1567 | |||
AK159304.1 | - | 10, 11 | 1586 | |||
AK213215.1 | - | 2, 3 | 96 | |||
BC033317.1 | - | 10, 11 | 1532 | 4459242 |