Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078649.1 | NP_001072117.1 | 3 | 339 | ||
NM_025889.2 | NP_080165.1 | 3 | 339 | |||
GenBank mRNA | AI789565.1 | - | 1 | 21 | ||
AK002220.1 | - | 3 | 333 | |||
AK008515.1 | - | 3 | 323 | |||
AK009263.1 | - | 3 | 341 | |||
AK009745.1 | - | 3 | 315 | |||
AK129014.1 | - | 3 | 312 | |||
AK166519.1 | - | 3 | 342 | |||
AK178280.1 | - | 1 | 12 | |||
AK195169.1 | - | 1 | 12 | |||
BC026765.1 | - | 3 | 321 | 4166287 | ||
CF983315.1 | - | 3 | 365 |