Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 10 | 1161 | ||
GenBank mRNA | AK005114.1 | - | 10 | 1195 | ||
AK007619.1 | - | 10 | 1206 | |||
AK007680.1 | - | 10 | 1135 | |||
AK008646.1 | - | 10 | 1160 | |||
AK009969.1 | - | 10 | 1164 | |||
AK017843.1 | - | 10 | 1178 | |||
AK150910.1 | - | 10 | 1184 | |||
AK151242.1 | - | 10 | 1162 | |||
AK152041.1 | - | 10 | 1184 | |||
AK152240.1 | - | 10 | 1193 | |||
AK160283.1 | - | 10 | 1184 | |||
AK162779.1 | - | 1162 | ||||
AK167236.1 | - | 10 | 1168 | |||
BC004694.1 | - | 10 | 1179 | 3498952 |