Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 8 | 923 | ||
GenBank mRNA | AK005114.1 | - | 8 | 957 | ||
AK007619.1 | - | 8 | 968 | |||
AK007680.1 | - | 8 | 897 | |||
AK009969.1 | - | 8 | 926 | |||
AK017843.1 | - | 8 | 940 | |||
AK150910.1 | - | 8 | 946 | |||
AK151242.1 | - | 8 | 924 | |||
AK152041.1 | - | 8 | 946 | |||
AK152240.1 | - | 8 | 955 | |||
AK160283.1 | - | 8 | 945 | |||
AK162779.1 | - | 924 | ||||
AK167236.1 | - | 8 | 931 | |||
BC004694.1 | - | 8 | 941 | 3498952 |