Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 11 | 1238 | ||
GenBank mRNA | AK005114.1 | - | 11 | 1272 | ||
AK007619.1 | - | 11 | 1283 | |||
AK007680.1 | - | 11 | 1212 | |||
AK008646.1 | - | 11 | 1237 | |||
AK009969.1 | - | 11 | 1241 | |||
AK017843.1 | - | 11 | 1255 | |||
AK150910.1 | - | 11 | 1261 | |||
AK151242.1 | - | 11 | 1239 | |||
AK152041.1 | - | 11 | 1261 | |||
AK152240.1 | - | 11 | 1270 | |||
AK160283.1 | - | 11 | 1261 | |||
AK167236.1 | - | 11 | 1245 | |||
BC004694.1 | - | 11 | 1256 | 3498952 |