Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025895.4 | NP_080171.1 | 1 | 230 | ||
GenBank mRNA | AF318060.1 | - | 1 | 154 | ||
AK003854.1 | - | 1 | 143 | |||
AK005681.1 | - | 1 | 139 | |||
AK007855.1 | - | 1 | 159 | |||
AK154249.1 | - | 1 | 156 | |||
BC024941.1 | - | 1 | 173 | 4017146 | ||
BY725533.1 | - | 1 | 210 | |||
DV653139.1 | - | 1 | 230 |