Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025899.2 | NP_080175.1 | 3, 4 | 365 | ||
GenBank mRNA | AK003975.1 | - | 3, 4 | 338 | ||
AK004492.1 | - | 3, 4 | 340 | |||
AK005151.1 | - | 3, 4 | 349 | |||
AK007669.1 | - | 2, 3 | 229 | |||
AK075856.1 | - | 3, 4 | 309 | |||
AK077583.1 | - | 3, 4 | 314 | |||
AK088103.1 | - | 3, 4 | 319 | |||
AK167377.1 | - | 3, 4 | 319 | |||
BC003423.1 | - | 3, 4 | 337 | 2609295 |