Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yae1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025904.3 | NP_080180.1 | 2 | 208 | ||
GenBank mRNA | AK005425.1 | - | 2 | 195 | ||
AK011774.1 | - | 2 | 207 | |||
AK011805.1 | - | 2 | 176 | |||
AK011852.1 | - | 2 | 176 | |||
AK019184.1 | - | 2 | 162 | |||
AK030455.1 | - | 1 | 1619 | |||
AK076034.1 | - | 2 | 165 | |||
AK151981.1 | - | 2 | 186 | |||
BC020123.1 | - | 2 | 202 | |||
BC037189.1 | - | 2 | 195 | |||
BC048453.1 | - | 2 | 191 | 6595060 | ||
BC083111.1 | - | 2 | 167 | 5329860 | ||
CJ084672.1 | - | 2 | 208 |