Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168475.1 | NP_001161947.1 | 6 | 1098 | ||
NM_001168476.1 | NP_001161948.1 | 5 | 898 | |||
NM_025905.3 | NP_080181.2 | 6 | 1098 | |||
GenBank mRNA | AK005431.1 | - | 5 | 859 | ||
AK076802.1 | - | 5 | 692 | |||
AK086049.1 | - | 1165 | ||||
AK141754.1 | - | 6 | 1084 | |||
BC038179.1 | - | 6 | 1069 | 4485073 |