Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025920.3 | NP_080196.3 | 4, 5 | 1797 | ||
GenBank mRNA | AK008304.1 | - | 2, 3 | 181 | ||
AK008587.1 | - | 2, 3 | 180 | |||
AK140888.1 | - | 1740 | ||||
AK167814.1 | - | 4, 5 | 1557 | |||
AK212297.1 | - | 1, 2 | 58 | |||
BC013538.1 | - | 3, 4 | 634 | |||
BC057963.1 | - | 3, 4 | 1095 | |||
BC063758.1 | - | 4, 5 | 1528 | 6334411 | ||
BC066042.1 | - | 4, 5 | 1732 | 6413779 |