Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 16, 17 | 2468 | ||
GenBank mRNA | AK011184.1 | - | 4, 5 | 823 | ||
AK076553.1 | - | 2, 3 | 596 | |||
AK087936.1 | - | 4, 5 | 793 | |||
AK142132.1 | - | 10, 11 | 1744 | |||
BC066150.1 | - | 5, 6 | 902 | |||
BC067400.1 | - | 5, 6 | 902 | |||
BC158104.1 | - | 15, 16 | 2348 | 9007388 |