Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024188.6 | NP_077150.1 | 5 | 855 | ||
GenBank mRNA | AB085609.1 | - | 5 | 597 | ||
AK009027.1 | - | 5 | 635 | |||
AK010790.1 | - | 5 | 598 | |||
AK011354.1 | - | 5 | 535 | |||
AK078086.1 | - | 5 | 855 | |||
AK146158.1 | - | 5 | 613 | |||
AK146342.1 | - | 5 | 600 | |||
AK151662.1 | - | 5 | 594 | |||
AK151695.1 | - | 5 | 585 | |||
AK167681.1 | - | 5 | 685 | |||
AK181182.1 | - | 1 | 71 | |||
AK181314.1 | - | 1 | 70 | |||
AK184597.1 | - | 1 | 71 | |||
BC003422.1 | - | 5 | 559 | 2598824 | ||
EF095212.1 | - | 2 | 248 |