Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005363.3 | NP_005354.1 | 3 | 957 | ||
NM_175868.2 | NP_787064.1 | 3 | 1021 | |||
GenBank mRNA | AK289437.1 | - | 3 | 941 | ||
BC041599.1 | - | 3 | 930 | 5458512 | ||
BC067731.1 | - | 3 | 938 | 4828429 | ||
BX640600.1 | - | 3 | 1318 | |||
D32076.1 | - | 1 | 815 | |||
EU161101.1 | - | 1 | 750 | |||
U10339.1 | - | 1 | 750 |
Gene Symbol: MAGEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR541768.1 | - | 1 | 750 | ||
CR542020.1 | - | 1 | 750 |