Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023294.2 | NP_075783.2 | 13 | 1610 | ||
GenBank mRNA | AF326730.1 | - | 13 | 1623 | ||
AK011497.1 | - | 13 | 1580 | |||
AK011554.1 | - | 9 | 1006 | |||
AK136236.1 | - | 12 | 1429 | |||
AK163317.1 | - | 13 | 1561 | |||
AK169427.1 | - | 13 | 1567 | |||
BC020131.1 | - | 13 | 1546 | 3709641 |
Gene Symbol: 2700099C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132724.1 | - | 7 | 821 | ||
BC002320.1 | - | 4 | 488 | |||
BC086941.1 | - | 2 | 105 |