Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025939.2 | NP_080215.1 | 2 | 478 | ||
GenBank mRNA | AK002669.1 | - | 2 | 139 | ||
AK010462.1 | - | 2 | 87 | |||
AK012118.1 | - | 2 | 236 | |||
AK013426.1 | - | 2 | 111 | |||
AK075949.1 | - | 2 | 108 | |||
AK079282.1 | - | 2 | 137 | |||
AK146722.1 | - | 3 | 430 | |||
AK159398.1 | - | 2 | 283 | |||
AK166264.1 | - | 2 | 133 | |||
BC018153.1 | - | 1 | 74 | 3586809 | ||
BC052691.1 | - | 2 | 478 | 5707686 |