Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024189.6 | NP_077151.3 | 4 | 529 | ||
GenBank mRNA | AB100452.1 | - | 4 | 519 | ||
AB100453.1 | - | 6 | 588 | |||
AK005008.1 | - | 4 | 513 | |||
AK011702.1 | - | 5 | 600 | |||
AK012787.1 | - | 4 | 435 | |||
AK148634.1 | - | 4 | 473 | |||
AK179353.1 | - | 2 | 226 | |||
AK184096.1 | - | 4 | 314 | |||
BC002192.1 | - | 4 | 402 | 3488691 |