Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024189.6 | NP_077151.3 | 3 | 427 | ||
GenBank mRNA | AB100452.1 | - | 3 | 417 | ||
AB100453.1 | - | 5 | 486 | |||
AK005008.1 | - | 3 | 411 | |||
AK011702.1 | - | 4 | 498 | |||
AK012787.1 | - | 3 | 333 | |||
AK148634.1 | - | 3 | 371 | |||
AK179353.1 | - | 1 | 124 | |||
AK184096.1 | - | 3 | 212 | |||
BC002192.1 | - | 3 | 300 | 3488691 |