Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082412.2 | NP_001075881.1 | 7 | 834 | ||
NM_001114179.1 | NP_001107651.1 | 2 | 216 | |||
GenBank mRNA | CX240158.1 | - | 2 | 216 |