Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175114.3 | NP_780323.2 | 3 | 472 | ||
GenBank mRNA | AK029123.1 | - | 4 | 870 | ||
AK041436.1 | - | 4 | 840 | |||
AK052264.1 | - | 4 | 859 | |||
BC058971.1 | - | 3 | 414 | 6832507 |