Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025969.4 | NP_080245.1 | 3 | 609 | ||
GenBank mRNA | AK006594.1 | - | 3 | 114 | ||
AK012203.1 | - | 3 | 609 | |||
AK013281.1 | - | 3 | 522 | |||
AK017477.1 | - | 3 | 577 | |||
AK029037.1 | - | 2 | 689 | |||
AK032468.1 | - | 3 | 610 | |||
AK168105.1 | - | 3 | 520 | |||
BC034297.1 | - | 3 | 584 | 4511762 |