Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290500.1 | NP_001277429.1 | 11 | 1755 | ||
NM_001290502.1 | NP_001277431.1 | 10 | 3791 | |||
NM_025978.4 | NP_080254.2 | 10 | 3553 | |||
NM_027619.4 | NP_081895.2 | 12 | 1911 | |||
GenBank mRNA | AK016428.1 | - | 1 | 386 | ||
AK038763.1 | - | 1 | 713 | |||
AK173315.1 | - | 11 | 1725 | |||
BC103802.1 | - | 1 | 370 |