Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025982.4 | NP_080258.1 | 4 | 420 | ||
GenBank mRNA | AK012457.1 | - | 4 | 421 | ||
AK012567.1 | - | 4 | 423 | |||
AK012853.1 | - | 4 | 423 | |||
AK013523.1 | - | 4 | 433 | |||
AK014183.1 | - | 4 | 419 | |||
AK075962.1 | - | 4 | 421 | |||
AK160269.1 | - | 2 | 1030 | |||
AK178759.1 | - | 3 | 158 | |||
AK180941.1 | - | 3 | 158 | |||
AK183488.1 | - | 3 | 136 | |||
AK188045.1 | - | 3 | 158 | |||
AK208118.1 | - | 3 | 134 | |||
BC023306.1 | - | 4 | 397 | 4165774 |