Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025990.4 | NP_080266.2 | 6 | 711 | ||
GenBank mRNA | AK009329.1 | - | 6 | 707 | ||
AK009441.1 | - | 6 | 713 | |||
AK009531.1 | - | 6 | 707 | |||
AK009562.1 | - | 6 | 707 | |||
AK009580.1 | - | 6 | 711 | |||
AK009629.1 | - | 6 | 710 | |||
AK009695.1 | - | 6 | 708 | |||
AK009803.1 | - | 6 | 709 | |||
AK009835.1 | - | 6 | 711 | |||
AK010012.1 | - | 6 | 711 | |||
AK010051.1 | - | 6 | 708 | |||
AK010115.1 | - | 6 | 707 | |||
AK010143.1 | - | 5 | 653 | |||
AK010145.1 | - | 6 | 711 | |||
AK010159.1 | - | 6 | 710 | |||
BC115526.1 | - | 6 | 690 | 40101263 | ||
BC115527.1 | - | 6 | 687 | 40101264 |