Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026010.2 | NP_080286.2 | 2 | 526 | ||
GenBank mRNA | AK008917.1 | - | 2 | 507 | ||
AK011925.1 | - | 2 | 526 | |||
AK013127.1 | - | 2 | 508 | |||
BC115472.1 | - | 2 | 526 | 40100448 | ||
BC115473.1 | - | 2 | 521 | 40100449 |