Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026011.3 | NP_080287.1 | 2 | 331 | ||
GenBank mRNA | AB185206.1 | - | 2 | 135 | ||
AK005999.1 | - | 2 | 178 | |||
AK009279.1 | - | 2 | 268 | |||
AK012024.1 | - | 2 | 295 | |||
AK030689.1 | - | 2 | 311 | |||
AK075941.1 | - | 2 | 292 | |||
AK076292.1 | - | 2 | 299 | |||
AK144177.1 | - | 2 | 302 | |||
AK146307.1 | - | 2 | 297 | |||
AK149822.1 | - | 2 | 281 | |||
AK152798.1 | - | 2 | 309 | |||
AK159348.1 | - | 2 | 297 | |||
AK169929.1 | - | 2 | 265 | |||
AK170645.1 | - | 2 | 297 | |||
AK195129.1 | - | 2 | 146 | |||
BC013719.1 | - | 2 | 280 | 3983809 | ||
BY261755.1 | - | 2 | 332 | |||
CT010337.1 | - | 2 | 135 |