Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026012.2 | NP_080288.1 | 3 | 355 | ||
GenBank mRNA | AF534394.1 | - | 2 | 167 | ||
AK007483.1 | - | 1 | 12 | |||
AK160222.1 | - | 3 | 341 | |||
BC039808.1 | - | 3 | 325 | 1547029 | ||
BC069856.1 | - | 3 | 355 | 6491606 |