Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025582.2 | NP_001020753.1 | 3 | 332 | ||
NM_001286986.1 | NP_001273915.1 | 3 | 329 | |||
NM_001286987.1 | NP_001273916.1 | 2 | 207 | |||
NM_026013.3 | NP_080289.1 | 3 | 332 | |||
GenBank mRNA | AI875295.1 | - | 3 | 332 | ||
AK008532.1 | - | 3 | 256 | |||
AK009940.1 | - | 3 | 269 | |||
AK012044.1 | - | 3 | 249 | |||
AK015888.1 | - | 3 | 295 | |||
AK052824.1 | - | 2 | 141 | |||
AK172455.1 | - | 3 | 299 | |||
BC024458.1 | - | 3 | 258 | 4975302 | ||
BI738450.1 | - | 3 | 300 |