Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025582.2 | NP_001020753.1 | 7 | 736 | ||
NM_001286986.1 | NP_001273915.1 | 9 | 994 | |||
NM_001286987.1 | NP_001273916.1 | 8 | 872 | |||
NM_026013.3 | NP_080289.1 | 9 | 997 | |||
GenBank mRNA | AK008532.1 | - | 7 | 660 | ||
AK009940.1 | - | 9 | 934 | |||
AK012044.1 | - | 9 | 914 | |||
AK015888.1 | - | 9 | 960 | |||
AK052824.1 | - | 8 | 806 | |||
AK172455.1 | - | 9 | 964 | |||
BC024458.1 | - | 7 | 662 | 4975302 |