Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026015.2 | NP_080291.1 | 2 | 189 | ||
GenBank mRNA | AK003361.1 | - | 2 | 182 | ||
AK007565.1 | - | 2 | 176 | |||
AK012098.1 | - | 2 | 190 | |||
AK013415.1 | - | 1 | 162 | |||
BC031796.1 | - | 2 | 168 | 4205598 |