Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026028.5 | NP_080304.1 | 12 | 1555 | ||
GenBank mRNA | AK012598.1 | - | 12 | 1447 | ||
AK031590.1 | - | 10 | 1359 | |||
BC018387.1 | - | 12 | 1383 | 3982552 | ||
CX205399.1 | - | 2 | 98 |