Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026028.5 | NP_080304.1 | 9 | 1075 | ||
GenBank mRNA | AK012598.1 | - | 9 | 967 | ||
BC018387.1 | - | 9 | 903 | 3982552 |