Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp11l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026031.3 | NP_080307.1 | 6 | 569 | ||
GenBank mRNA | AK007361.1 | - | 4 | 340 | ||
AK008801.1 | - | 2 | 121 | |||
AK012550.1 | - | 6 | 570 | |||
AK145266.1 | - | 6 | 602 | |||
AK147113.1 | - | 6 | 600 | |||
BC043139.1 | - | 5 | 1277 | |||
BC061074.1 | - | 6 | 598 | 6706808 |