Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp11l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026031.3 | NP_080307.1 | 3 | 247 | ||
GenBank mRNA | AK007361.1 | - | 1 | 18 | ||
AK012550.1 | - | 3 | 248 | |||
AK084149.1 | - | 1 | 586 | |||
AK145266.1 | - | 3 | 280 | |||
AK147113.1 | - | 3 | 278 | |||
BC043139.1 | - | 3 | 227 | |||
BC061074.1 | - | 3 | 276 | 6706808 |