Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026034.4 | NP_080310.1 | 5 | 745 | ||
GenBank mRNA | AK009821.1 | - | 5 | 739 | ||
AK011303.1 | - | 5 | 744 | |||
AK012872.1 | - | 5 | 720 | |||
AK015272.1 | - | 5 | 878 | |||
BC027343.1 | - | 5 | 676 | |||
BC038487.1 | - | 5 | 697 | 2812558 | ||
BC046281.1 | - | 5 | 697 | 6395384 | ||
BC058573.1 | - | 5 | 786 | 6487440 |