Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026034.4 | NP_080310.1 | 1, 2 | 251 | ||
GenBank mRNA | AK009821.1 | - | 1, 2 | 244 | ||
AK011303.1 | - | 1, 2 | 250 | |||
AK012872.1 | - | 1, 2 | 226 | |||
BC038487.1 | - | 1, 2 | 203 | 2812558 | ||
BC046281.1 | - | 1, 2 | 203 | 6395384 | ||
BC058573.1 | - | 1, 2 | 292 | 6487440 |