Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026040.3 | NP_080316.3 | 5 | 395 | ||
GenBank mRNA | AK012865.1 | - | 5 | 397 | ||
AK141882.1 | - | 389 | ||||
AK144845.1 | - | 394 | ||||
AK147692.1 | - | 373 | ||||
AY611629.1 | - | 5 | 368 | |||
BC024338.1 | - | 372 | 5009612 |