Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026041.2 | NP_080317.3 | 2 | 346 | ||
GenBank mRNA | AK010658.1 | - | 2 | 314 | ||
AK013204.1 | - | 2 | 324 | |||
AK013217.1 | - | 2 | 314 | |||
AK087997.1 | - | 2 | 316 | |||
AK169088.1 | - | 2 | 347 | |||
BC006787.1 | - | 2 | 312 | 3589453 |